L(+)-Rhamnose Monohydrat
Lieferant: Apollo Scientific
|
Achtung
|
BIK9015-1KG
BIK9015-100G
BIK9015-500G
APOSBIK9015-1KGEA
237
EUR
APOSBIK9015-1KG
APOSBIK9015-100G
APOSBIK9015-500G
L(+)-Rhamnose Monohydrat
L(+)-Rhamnose Monohydrat
Formel:
C₆H₁₂O₅·1H₂O Schmelzpunkt: 90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149363 CAS-Nummer: 10030-85-0 EINECS: 222-793-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...