Lestaurtinib

Lieferant: Apollo Scientific
Achtung

BIL2000-1MG
APOSBIL2000-1MGEA 244 EUR
APOSBIL2000-1MG
Lestaurtinib
Lestaurtinib

  • Protein

Lagertemperatur: Gefrierschrank
CAS-Nummer: 111358-88-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR