HRP-conjugate stabiliser, SeramunStab® STA2
Lieferant: Apollo Scientific
BIL3112-100ML
APOSBIL3112-100MLEA 50.9 EUR
APOSBIL3112-100ML
HRP-conjugate stabiliser, SeramunStab® STA2
Proteinkonjugate und Markierungen
Liquid product red odourless pH-value 7,3 to 7,5 Boiling point 102 °C. Stabiliser for a broad range of HRP conjugates.