HRP-conjugate stabiliser, SeramunStab® STNP
Lieferant: Apollo Scientific
BIL3113-100ML
APOSBIL3113-100MLEA 62.5 EUR
APOSBIL3113-100ML
HRP-conjugate stabiliser, SeramunStab® STNP
Proteinkonjugate und Markierungen
Liquid product red odourless pH 6,7 to 6,9. Protein free Stabiliser. Enzyme activity preservation by special chemistry.