SeramunBlock
Lieferant: Apollo Scientific
BIL3122-100ML BIL3122-250ML
APOSBIL3122-100MLEA 0 EUR
APOSBIL3122-100ML APOSBIL3122-250ML
SeramunBlock
Proteinkonjugate und Markierungen
Combined Stabiliser/ blocking solution for solid phase immobilised proteins effecting protein stabilisation and blocking of free surfaces.