Lactulose
Lieferant: Apollo Scientific
|
Achtung
|
BIL9700-100G
BIL9700-250G
BIL9700-500G
APOSBIL9700-100GEA
277
EUR
APOSBIL9700-100G
APOSBIL9700-250G
APOSBIL9700-500G
Lactulose
Lactulose
Formel:
C₁₂H₂₂O₁₁ Schmelzpunkt: 169 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00151469 CAS-Nummer: 4618-18-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...