6-Mercaptopurine monohydrate

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Purinethiol monohydrate, 6-Mercaptopurine hydrate, 7H-Purine-6-thiol hydrate, 6-Thiohypoxanthine monohydrate

BIM0129-25G BIM0129-5G
APOSBIM0129-25GEA 46.4 EUR
APOSBIM0129-25G APOSBIM0129-5G
6-Mercaptopurine monohydrate
6-Mercaptopurine monohydrate

6-Mercaptopurine is an antineoplastic agent which acts asan antimetabolite. It is an analogue of the natural purines hypoxanthine and adenine.

Formel: C₅H₄N₄S·H₂O
Schmelzpunkt: 315 °C (Zersetzung)
Lagertemperatur: Kühlschrank
MDL: MFCD03854445
CAS-Nummer: 6112-76-1
EINECS: 200-037-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR