MOPS-Na ≥98%
Lieferant: Apollo Scientific
Synonyme:
Natrium-4-morpholin-1-ylpropylsulfonat, MOPS Natriumsalz, 3-(N-Morpholino)propansulfonsäure Natriumsalz
BIM0197-500G
APOSBIM0197-500GEA
89.9
EUR
APOSBIM0197-500G
MOPS-Na ≥98%
MOPS-Na
- Buffer
Formel:
C₇H₁₄NNaO₄S Molecular Weight: 231,25 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064350 CAS-Nummer: 71119-22-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...