Agar
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Agar Agar
BIM1002-1KG
BIM1002-500G
BIM1002-5KG
APOSBIM1002-1KGEA
125
EUR
APOSBIM1002-1KG
APOSBIM1002-500G
APOSBIM1002-5KG
Agar
Agar
A purified agar with a high gel strength and clarity for tissue culture and microbiological work. Minimal conc. of 5.0g/L to obtain solid gel.
Schmelzpunkt:
90 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00081288 CAS-Nummer: 9002-18-0 EINECS: 232-658-1 Merck Index: 13,00184 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...