Agar

Lieferant: Apollo Scientific
Achtung

Synonyme: Agar Agar

BIM1002-1KG BIM1002-500G BIM1002-5KG
APOSBIM1002-1KGEA 125 EUR
APOSBIM1002-1KG APOSBIM1002-500G APOSBIM1002-5KG
Agar
Agar

A purified agar with a high gel strength and clarity for tissue culture and microbiological work. Minimal conc. of 5.0g/L to obtain solid gel.

Schmelzpunkt: 90
Lagertemperatur: Raumtemperatur
MDL: MFCD00081288
CAS-Nummer: 9002-18-0
EINECS: 232-658-1
Merck Index: 13,00184

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR