3-Maleimidopropionic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)propanoic acid, 3-Maleimido-propionic acid, N-(2-Carboxyethyl)maleimide, MPS-Acid, N-Maleoyl-β-alanine
BIM1007-1G
APOSBIM1007-1GEA
21.9
EUR
APOSBIM1007-1G
3-Maleimidopropionic acid
3-Maleimidopropionic acid
Protein modifier.
Formel:
C₇H₇NO₄ Molecular Weight: 169,14 g/mol Schmelzpunkt: 103…106 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00043030 CAS-Nummer: 7423-55-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...