Maltooctaose
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
FT-0670933, (Hex)8, (2R,3R,5R)-4-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal, (2R,3R,5R)-4-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-5-(((2R,3R,4R,5S,6R)-3,4-Dihydroxy-6-(hydroxymethyl)-5-(((2R,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-2,3,5,6-tetrahydroxyhexanal, G8
Formel:
C₄₈H₈₂O₄₁ Molecular Weight: 1315.14 g/mol Schmelzpunkt: 212 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD05865015 CAS-Nummer: 66567-45-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...