4-Hydroxyphenyl glyoxal (HPG)
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
HPG
BIM104-100MG
APOSBIM104-100MGEA
270
EUR
APOSBIM104-100MG
4-Hydroxyphenyl glyoxal (HPG)
4-Hydroxyphenyl glyoxal (HPG)
Modifies arginine residues
- Protein
Ref: Yamasaki,R.B,Vega,A.and Freeney,R.E.(1980)Anal.Biochem.109, 32-40
Formel:
C₈H₆O₃ Molecular Weight: 150,131 g/mol Schmelzpunkt: 106…112 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00054984 CAS-Nummer: 24645-80-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...