Sulphosuccinimidyl-4-[2-(4,4-dimethoxytrityl)]butyrate

Lieferant: Apollo Scientific
Achtung

Synonyme: Sulpho SDTB

BIM109-50MG
APOSBIM109-50MGEA 531 EUR
APOSBIM109-50MG
Sulphosuccinimidyl-4-[2-(4,4-dimethoxytrityl)]butyrate
Sulphosuccinimidyl-4-[2-(4,4-dimethoxytrityl)]butyrate

Detects immobilized amines


  • Protein


Ref: Gaur.R.K. and Gupta,K.C. (1989) Anal.Biochem. 180, 253-258

Formel: C₂₉H₂₉NO₁₀S
Molecular Weight: 583.606 g/mol
Schmelzpunkt: 114…116 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00153609
CAS-Nummer: 199436-86-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR