4-Methylumbelliferyl-Α-L-arabinopyranoside

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Methylumbelliferyl alpha-L-Arabinopyranoside

BIM1090-100MG
APOSBIM1090-100MGEA 36.4 EUR
APOSBIM1090-100MG
4-Methylumbelliferyl-Α-L-arabinopyranoside
4-Methylumbelliferyl-Α-L-arabinopyranoside
Formel: C₁₅H₁₆O₇
Molecular Weight: 308,28 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00064087
CAS-Nummer: 69414-26-2
EINECS: 273-991-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR