4-Methylumbelliferyl-α-L-fucoside
Lieferant: Apollo Scientific
|
Achtung
|
BIM1093-25MG
APOSBIM1093-25MGEA
248
EUR
APOSBIM1093-25MG
4-Methylumbelliferyl-α-L-fucoside
4-Methylumbelliferyl-α-L-fucoside
Formel:
C₁₆H₁₈O₇ Molecular Weight: 322,313 g/mol Schmelzpunkt: 230 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD00051218 CAS-Nummer: 54322-38-2 EINECS: 259-098-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...