N-(2-Iodethyl)-2,2,2-trifluoracetamid

Lieferant: Apollo Scientific
Achtung

BIM110-1G
APOSBIM110-1GEA 161 EUR
APOSBIM110-1G
N-(2-Iodethyl)-2,2,2-trifluoracetamid
N-(2-Iodethyl)-2,2,2-trifluoracetamid

Modifies sulphhydryl on cysteine. Substitute for ethylene imine.

Formel: C₄H₅F₃INO
Lagertemperatur: Kühlschrank
CAS-Nummer: 67680-56-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR