Diethylenetriaminepentaacetic dianhydride

Lieferant: Apollo Scientific
Achtung

BIM112-1G
APOSBIM112-1GEA 0 EUR
APOSBIM112-1G
Diethylenetriaminepentaacetic dianhydride
Diethylenetriaminepentaacetic dianhydride

Introduces chelator into proteins for radiolabelingRef: Hnatowich,D.J,et al.(1980) Science 220, 613-615Ref: Layne,W.W,et al.(1982) J.Nucl.Med. 23, 627-630

Formel: C₁₄H₁₉N₃O₈
Schmelzpunkt: 182…184 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010697
CAS-Nummer: 23911-26-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR