4-Nitrophenyl-α-D-mannopyranosid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Nitrophenyl α-D-Mannopyranoside, 4-Nitrophenyl a-D-mannopyranoside

BIM1208-1G BIM1208-500MG
APOSBIM1208-1GEA 59.6 EUR
APOSBIM1208-1G APOSBIM1208-500MG
4-Nitrophenyl-α-D-mannopyranosid
4-Nitrophenyl-α-D-mannopyranosid
Formel: C₁₂H₁₅NO₈
Schmelzpunkt: 180 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00066002
CAS-Nummer: 10357-27-4
EINECS: 233-776-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR