4-Methylumbelliferyl caprylate

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Methyl-2-oxo-2H-chromen-7-yl octanoate

BIM2042-250MG
APOSBIM2042-250MGEA 31.9 EUR
APOSBIM2042-250MG
4-Methylumbelliferyl caprylate
4-Methylumbelliferyl caprylate
Formel: C₁₈H₂₂O₄
Molecular Weight: 302,365 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00038659
CAS-Nummer: 20671-66-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR