4-Methylumbelliferyl β-D-galactopyranoside-6-sulphate sodium salt
Lieferant: Apollo Scientific
|
Achtung
|
BIM2043-25MG
APOSBIM2043-25MGEA
0
EUR
APOSBIM2043-25MG
4-Methylumbelliferyl β-D-galactopyranoside-6-sulphate sodium salt
4-Methylumbelliferyl β-D-galactopyranoside-6-sulphate sodium salt
Formel:
C₁₆H₁₇O₁₁S·Na Schmelzpunkt: 158…160 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD01861961 CAS-Nummer: 206443-06-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...