4-Methylumbelliferyl α-L-iduronide (free acid)
Lieferant: Apollo Scientific
|
Achtung
|
BIM2044-2MG
APOSBIM2044-2MGEA
226
EUR
APOSBIM2044-2MG
4-Methylumbelliferyl α-L-iduronide (free acid)
4-Methylumbelliferyl α-L-iduronide (free acid)
Lagertemperatur:
Gefrierschrank |
MDL:
MFCD00135894 CAS-Nummer: 66966-09-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...