4-Methylumbelliferyl beta-D-ribofuranoside

Lieferant: Apollo Scientific
Achtung

BIM2050-50MG
APOSBIM2050-50MGEA 265 EUR
APOSBIM2050-50MG
4-Methylumbelliferyl beta-D-ribofuranoside
4-Methylumbelliferyl beta-D-ribofuranoside
Formel: C₁₅H₁₆O₇
Lagertemperatur: Gefrierschrank
MDL: MFCD06796941
CAS-Nummer: 195385-93-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR