IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥99% dioxanfrei

Lieferant: Apollo Scientific
Achtung

Synonyme: IPTG

BIMB1008-25G BIMB1008-5G BIMB1008-10G
APOSBIMB1008-25GEA 87 EUR
APOSBIMB1008-25G APOSBIMB1008-5G APOSBIMB1008-10G
IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥99% dioxanfrei
IPTG (Isopropyl-β-D-thiogalactopyranosid)

IPTG is used in conjunction with X-Gal for detection of lac+ colonies or cells in a colorimetric assay to detect recombinants (white) from non-recombinants (blue) in cloning strategies.

Formel: C₉H₁₈O₅S
Molecular Weight: 238.31 g/mol
Siedepunkt: 402
Schmelzpunkt: 105 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00063273
CAS-Nummer: 367-93-1
EINECS: 206-703-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR