X-Gluc (5-Brom-4-chlor-3-indolyl-ß-D-glucuronid Cyclohexylammoniumsalz, X-GlcA) ≥98%

Lieferant: Apollo Scientific
Achtung

BIMB1021-1G BIMB1021-250MG BIMB1021-500MG
APOSBIMB1021-1GEA 0 EUR
APOSBIMB1021-1G APOSBIMB1021-250MG APOSBIMB1021-500MG
X-Gluc (5-Brom-4-chlor-3-indolyl-ß-D-glucuronid Cyclohexylammoniumsalz, X-GlcA) ≥98%
X-Gluc (5-Brom-4-chlor-3-indolyl-ß-D-glucuronid Cyclohexylammoniumsalz, X-GlcA)

X-GlcA is a substrate for b-Glucuronidase (GUS) encoded by the gusA gene. The substrate is used as a qualitative marker of specific GUS expression.

Formel: C₂₀H₂₆BrClN₂O₇
Lagertemperatur: Kühlschrank
MDL: MFCD00058543
CAS-Nummer: 114162-64-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR