5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt ≥98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
XGLUC Sodium Salt, 5-Bromo-4-chloro-3-indolyl-b-D-glucuronide sodium salt
BIMB1121-1G
BIMB1121-250MG
BIMB1121-500MG
APOSBIMB1121-1GEA
114
EUR
APOSBIMB1121-1G
APOSBIMB1121-250MG
APOSBIMB1121-500MG
5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt ≥98%
5-Bromo-4-chloro-3-indolyl-beta-D-glucuronide sodium salt
Histochemical substrate for b-Glucuronidase.Produces blue-black water insoluble colour.
Formel:
C₁₄H₁₃BrClNO₇ Lagertemperatur: Kühlschrank |
MDL:
MFCD00135782 CAS-Nummer: 129541-41-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...