5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt ≥98%

Lieferant: Apollo Scientific
Achtung

Synonyme: XGLUC Sodium Salt, 5-Bromo-4-chloro-3-indolyl-b-D-glucuronide sodium salt

BIMB1121-1G BIMB1121-250MG BIMB1121-500MG
APOSBIMB1121-1GEA 114 EUR
APOSBIMB1121-1G APOSBIMB1121-250MG APOSBIMB1121-500MG
5-Bromo-4-chloro-3-indolyl-β-D-glucuronide sodium salt ≥98%
5-Bromo-4-chloro-3-indolyl-beta-D-glucuronide sodium salt

Histochemical substrate for b-Glucuronidase.Produces blue-black water insoluble colour.

Formel: C₁₄H₁₃BrClNO₇
Lagertemperatur: Kühlschrank
MDL: MFCD00135782
CAS-Nummer: 129541-41-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR