Octyl-β-D-thioglucopyranosid ≥99%, ultrarein

Lieferant: Apollo Scientific
Achtung

BIMB1375-1G BIMB1375-5G BIMB1375-25G
APOSBIMB1375-1GEA 21.9 EUR
APOSBIMB1375-1G APOSBIMB1375-5G APOSBIMB1375-25G
Octyl-β-D-thioglucopyranosid ≥99%, ultrarein
Octyl-β-D-thioglucopyranosid

Non-ionic detergent used for membrane protein solubisation.

Formel: C₁₄H₂₈O₅S
Schmelzpunkt: 125…131 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00012189
CAS-Nummer: 85618-21-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR