Octyl-β-D-thioglucopyranosid ≥99%, ultrarein
Lieferant: Apollo Scientific
|
Achtung
|
BIMB1375-1G
BIMB1375-5G
BIMB1375-25G
APOSBIMB1375-1GEA
21.9
EUR
APOSBIMB1375-1G
APOSBIMB1375-5G
APOSBIMB1375-25G
Octyl-β-D-thioglucopyranosid ≥99%, ultrarein
Octyl-β-D-thioglucopyranosid
Non-ionic detergent used for membrane protein solubisation.
Formel:
C₁₄H₂₈O₅S Schmelzpunkt: 125…131 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00012189 CAS-Nummer: 85618-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...