IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥99% tier-, dioxanfrei

Lieferant: Apollo Scientific
Achtung

Synonyme: IPTG

BIMB8001-25G BIMB8001-5G
APOSBIMB8001-25GEA 109 EUR
APOSBIMB8001-25G APOSBIMB8001-5G
IPTG (Isopropyl-β-D-thiogalactopyranosid) ≥99% tier-, dioxanfrei
IPTG (Isopropyl-β-D-thiogalactopyranosid)

Material is synthesised from animal-free material.

Formel: C₉H₁₈O₅S
Molecular Weight: 238.31 g/mol
Siedepunkt: 402
Schmelzpunkt: 109…115 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00063273
CAS-Nummer: 367-93-1
EINECS: 206-703-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR