Cladribin

Lieferant: Apollo Scientific
Gefahr

BIMN2008-100MG
APOSBIMN2008-100MGEA 46.4 EUR
APOSBIMN2008-100MG
Cladribin
Cladribine
Formel: C₁₀H₁₂ClN₅O₃
Lagertemperatur: Kühlschrank
MDL: MFCD00153939
CAS-Nummer: 4291-63-8
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR