3’- Amino-3’-deoxyadenosine

Lieferant: Apollo Scientific

BIMN2044-100MG
APOSBIMN2044-100MGEA 548 EUR
APOSBIMN2044-100MG
3’- Amino-3’-deoxyadenosine
3’- Amino-3’-deoxyadenosine
Formel: C10H14N6O3
Molecular Weight: 266,26 g/mol
Lagertemperatur: Raumtemperatur
CAS-Nummer: 2504-55-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR