Nonactin (aus Streptomyces griseus)

Lieferant: Apollo Scientific
Achtung

Synonyme: Polynactin, Werramycin A

BIN0110-10MG BIN0110-25MG
APOSBIN0110-10MGEA 102 EUR
APOSBIN0110-10MG APOSBIN0110-25MG
Nonactin (aus Streptomyces griseus)
Nonactin
Formel: C₄₀H₆₄O₁₂
Molecular Weight: 736.9 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 6833-84-7
EINECS: 229-911-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR