Nigericin sodium salt

Lieferant: Apollo Scientific
Gefahr

BIN0112-10MG BIN0112-25MG
APOSBIN0112-10MGEA 116 EUR
APOSBIN0112-10MG APOSBIN0112-25MG
Nigericin sodium salt
Nigericin sodium salt
Formel: C₄₀H₆₇NaO₁₁
Lagertemperatur: Kühlschrank
MDL: MFCD00036825
CAS-Nummer: 28643-80-3
UN: 3462
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR