Neomycinsulfat

Lieferant: Apollo Scientific
Gefahr

BIN0135-100G BIN0135-25G
APOSBIN0135-100GEA 84.1 EUR
APOSBIN0135-100G APOSBIN0135-25G
Neomycinsulfat
Neomycinsulfat

Neomycin is an aminoglycoside antibiotic and has a bactericidal action against many gram-negative bacteria.


  • Potency: 710mg/mg
  • Neomycin B: 85%
  • Neomycin C: 10%
  • Cell culture tested

Formel: C₂₃H₄₆N₆O₁₃·3H₂SO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00150827
CAS-Nummer: 1405-10-3
EINECS: 215-773-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR