Naphthol AS-TR-phosphat Dinatriumsalz

Lieferant: Apollo Scientific
Achtung

BIN0365-1G BIN0365-500MG BIN0365-1680G BIN0365-240G BIN0365-480G
APOSBIN0365-1GEA 306 EUR
APOSBIN0365-1G APOSBIN0365-500MG APOSBIN0365-1680G APOSBIN0365-240G APOSBIN0365-480G
Naphthol AS-TR-phosphat Dinatriumsalz
Naphthol AS-TR-phosphat Dinatriumsalz

Substrate for the histochemical demonstration of acid and alkaline phosphatase

Formel: C₁₈H₁₃ClNNa₂O₅P
Lagertemperatur: Gefrierschrank
CAS-Nummer: 4264-93-1

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR