p-Nitrophenyl phosphate DiTris salt

Lieferant: Apollo Scientific
Achtung

BIN4001-100G BIN4001-25G BIN4001-5G
APOSBIN4001-100GEA 284 EUR
APOSBIN4001-100G APOSBIN4001-25G APOSBIN4001-5G
p-Nitrophenyl phosphate DiTris salt
p-Nitrophenyl phosphate DiTris salt

Enzyme substrate for determination of phosphatase.

Formel: C₆H₆NO₆P
Lagertemperatur: Kühlschrank
MDL: MFCD00037009
CAS-Nummer: 68189-42-4
EINECS: 269-198-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR