Penicillin G Natriumsalz

Lieferant: Apollo Scientific
Gefahr

BIP0142-100G BIP0142-250G BIP0142-25G
APOSBIP0142-100GEA 0 EUR
APOSBIP0142-100G APOSBIP0142-250G APOSBIP0142-25G
Penicillin G Natriumsalz
Penicillin G Natriumsalz

Penicillin G is an inhibitor of bacterial cell wall synthesis.Active against gram-positive and some gram-negative bacteria. b-lactamase sensitive.


  • Potency: 1670 mg/mg

Formel: C₁₆H₁₇N₂NaO₄S
Lagertemperatur: Kühlschrank
MDL: MFCD00069666
CAS-Nummer: 69-57-8
EINECS: 200-710-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR