Polymyxin B-Sulfat

Lieferant: Apollo Scientific
Achtung

BIP0145-10G BIP0145-1G BIP0145-5G
APOSBIP0145-10GEA 219 EUR
APOSBIP0145-10G APOSBIP0145-1G APOSBIP0145-5G
Polymyxin B-Sulfat
Polymyxin B sulphate

Poly B is a mixture of the sulphates of polypeptides


  • approx. 8000 units/mg
  • Cell culture tested

Formel: C₅₅H₉₆N₁₆O₁₃·2H₂SO₄
Lagertemperatur: Kühlschrank
MDL: MFCD00131991
CAS-Nummer: 1405-20-5
EINECS: 215-774-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR