Phosphoenolpyruvic acid monocyclohexylammonium salt
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
PEP·CHX
BIP0688-1G
APOSBIP0688-1GEA
21.9
EUR
APOSBIP0688-1G
Phosphoenolpyruvic acid monocyclohexylammonium salt
Phosphoenolpyruvic acid monocyclohexylammonium salt
Formel:
C₃H₅O₆P Molecular Weight: 267,22 g/mol Lagertemperatur: Gefrierschrank |
MDL:
MFCD00036375 CAS-Nummer: 10526-80-4 EINECS: 234-084-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...