L-α-Phenylalanin ≥99,5%

Lieferant: Apollo Scientific
Achtung

Synonyme: H-Phe-OH, H-L-Phe-OH, L-α-Phenylalanin, 3-Phenyl-L(-)-alanin, 3-Phenyl-L-alanin, (S)-(-)-2-Amino-3-phenylpropansäure

BIP0716-100G BIP0716-1KG BIP0716-500G
APOSBIP0716-100GEA 21.9 EUR
APOSBIP0716-100G APOSBIP0716-1KG APOSBIP0716-500G
L-α-Phenylalanin ≥99,5%
L-α-Phenylalanin

USP, Pyrogen free

Formel: C₉H₁₁NO₂
Schmelzpunkt: 270…275 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064227
CAS-Nummer: 63-91-2
EINECS: 200-568-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR