Phalloidin

Lieferant: Apollo Scientific
Gefahr

BIP0842-1MG
APOSBIP0842-1MGEA 306 EUR
APOSBIP0842-1MG
Phalloidin
Phalloidin

Interferes with function of actin rich structures bybinding to polymeric F actin

Formel: C₃₅H₄₈N₈O₁₁S
Molecular Weight: 788,9 g/mol
Lagertemperatur: Kühlschrank
CAS-Nummer: 17466-45-4
EINECS: 241-484-5
UN: 3462
ADR: 6.1,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR