Perillartine 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: (NZ)-N-[(4-prop-1-en-2-ylcyclohexen-1-yl)methylidene]hydroxylamine, (E)-4-(Prop-1-en-2-yl)cyclohex-1-enecarbaldehyde oxime

BIP4001-25G
APOSBIP4001-25GEA 56.7 EUR
APOSBIP4001-25G
Perillartine 95%
Perillartine
Formel: C₁₀H₁₅NO
Molecular Weight: 165,232 g/mol
Schmelzpunkt: 101 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00019421
CAS-Nummer: 30950-27-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR