Sulpho-SASD (Sulphosuccinimidyl 2-(4-azidosalicylamido)ethyl-1,3-dithiopropionate)
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Sulfo SASD
BIPA102-100MG
APOSBIPA102-100MGEA
212
EUR
APOSBIPA102-100MG
Sulpho-SASD (Sulphosuccinimidyl 2-(4-azidosalicylamido)ethyl-1,3-dithiopropionate)
Sulpho-SASD (Sulphosuccinimidyl 2-(4-azidosalicylamido)ethyl-1,3-dithiopropionate)
Photoreactive -Primary amine reactive Iodinatable -Water soluble
Ref: Ji, T.H and Ji I.(1982)Anal.Biochem.121, 286-289
Formel:
C₁₆H₁₆N₅NaO₉S₃ Molecular Weight: 541,51 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861963 CAS-Nummer: 144650-95-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...