Bis[2-(4-azidosalicylamido)ethyl]disulphide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
BASED
BIPA104-100MG
APOSBIPA104-100MGEA
435
EUR
APOSBIPA104-100MG
Bis[2-(4-azidosalicylamido)ethyl]disulphide
Bis[2-(4-azidosalicylamido)ethyl]disulphide
Photoreactive -Primary amine reactive -Iodinatable
Formel:
C₁₈H₁₈N₈O₄S₂ Molecular Weight: 474,51 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861964 CAS-Nummer: 199804-21-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...