N-[4-(4-Azidosalicylamido)butyl]3-(2'-pyridyldithio)propionamide
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
APDP
BIPA105-100MG
APOSBIPA105-100MGEA
145
EUR
APOSBIPA105-100MG
N-[4-(4-Azidosalicylamido)butyl]3-(2'-pyridyldithio)propionamide
N-[4-(4-Azidosalicylamido)butyl]3-(2'-pyridyldithio)propionamide
Photoreactive -Sulphhydryl reactive -Cleavable -Iodinatable
Ref: Traut,R.R ,et al.Protein Function, A Practical Approach.Editedby T.E. Creighton.IRL Press, 1989, p.101
Formel:
C₁₉H₂₂N₆O₃S₂ Molecular Weight: 446,546 g/mol Schmelzpunkt: 107 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00078542 CAS-Nummer: 147492-84-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...