4-Azidophenyl glyoxal hydrate

Lieferant: Apollo Scientific
Achtung

Synonyme: APG

BIPA108-100MG
APOSBIPA108-100MGEA 125 EUR
APOSBIPA108-100MG
4-Azidophenyl glyoxal hydrate
4-Azidophenyl glyoxal hydrate

Photoreactive -Arginine residue reactive


Ref: Ngo,T.T,et al(1981)J.Biol.Chem.256, 11313-11318Ref: Sgro,J, et al(1986)Eur.J.Biochem.154, 69-76

Formel: C₈H₅N₃O₂·H₂O
Molecular Weight: 193,159 g/mol
Schmelzpunkt: 95…122 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00150595
CAS-Nummer: 1196151-49-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR