Sulpho-SAND (Sulphosuccinimidyl 2-(3-azido-2-nitrobenzamido)ethyl-1,3'-dithiopropionate)
Lieferant: Apollo Scientific
Synonyme:
Sulfo SAND
BIPA111-100MG
APOSBIPA111-100MGEA
174
EUR
APOSBIPA111-100MG
Sulpho-SAND (Sulphosuccinimidyl 2-(3-azido-2-nitrobenzamido)ethyl-1,3'-dithiopropionate)
Sulpho-SAND (Sulphosuccinimidyl 2-(3-azido-2-nitrobenzamido)ethyl-1,3'-dithiopropionate)
Photoreactive -Primary amine reactive Thiol cleavable -Water soluble
Ref: Lewis,R.V,et al(1977) Biochemistry 16, 5650-5654