Sulpho-SADP (Sulphosuccinimidyl 4-azidophenyldithiopropionate)
Lieferant: Apollo Scientific
Synonyme: Sulfo SADP
BIPA112-100MG
APOSBIPA112-100MGEA 145 EUR
APOSBIPA112-100MG
Sulpho-SADP (Sulphosuccinimidyl 4-azidophenyldithiopropionate)
Sulpho-SADP (Sulphosuccinimidyl 4-azidophenyldithiopropionate)
Photoreactive -Primary amine reactive Cleavable -Water soluble
Ref: Zarling,D.A,et al(1982)J.Immunol. 128, 251-257