Sulpho-SAED (Sulphosuccinimidyl 2-(7-azido-4-methylcoumarin-3-acetamide)ethyl-1,3-dithiopropionate)
Lieferant: Apollo Scientific
Achtung
Synonyme: Sulfo SAED
BIPA113-50MG
APOSBIPA113-50MGEA 219 EUR
APOSBIPA113-50MG
Sulpho-SAED (Sulphosuccinimidyl 2-(7-azido-4-methylcoumarin-3-acetamide)ethyl-1,3-dithiopropionate)
Sulpho-SAED (Sulphosuccinimidyl 2-(7-azido-4-methylcoumarin-3-acetamide)ethyl-1,3-dithiopropionate)
Photoreactive -Primary amine reactive -Fluorescent label
Ref: Kang,J.J, et al (1991) Biophys.J.59, 249aRef: Thevenin,B.J,et al(1991)Biophys.J.59, 358a